Product Details

SNP ID
rs200725819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123940066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCCTTATGGCAGCCTCCTGGGT[A/G]GGGGGCTTCATCCACTCCATAGTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D5 PubMed Links

Gene Details

Gene
OR4D5
Gene Name
olfactory receptor family 4 subfamily D member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001965.1 450 Silent Mutation GTA,GTG V150V NP_001001965.1
Gene
OR6T1
Gene Name
olfactory receptor family 6 subfamily T member 1
There are no transcripts associated with this gene.

View Full Product Details