Product Details

SNP ID
rs200916709
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117181514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGAGGCTGTGGTGTCCTTCCA[A/G]GTGCCCCTAATCCTGCGAGGGATGT
Phenotype
MIM: 602508
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PAFAH1B2 PubMed Links

Gene Details

Gene
PAFAH1B2
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 2
There are no transcripts associated with this gene.

Gene
SIDT2
Gene Name
SID1 transmembrane family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040455.1 323 Silent Mutation CAA,CAG Q94Q NP_001035545.1

View Full Product Details