Product Details

SNP ID
rs200710595
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62672088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAACTGGATGGTTGGCGGCGAA[A/G]GCGGCGCCGGCGGGAGGTCACCGTG
Phenotype
MIM: 616378 MIM: 616097
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C11orf98 PubMed Links

Gene Details

Gene
C11orf98
Gene Name
chromosome 11 open reading frame 98
There are no transcripts associated with this gene.

Gene
LBHD1
Gene Name
LBH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024099.3 348 Intron NP_077004.2
Gene
METTL12
Gene Name
methyltransferase like 12
There are no transcripts associated with this gene.

Gene
SNORA57
Gene Name
small nucleolar RNA, H/ACA box 57
There are no transcripts associated with this gene.

Gene
UBXN1
Gene Name
UBX domain protein 1
There are no transcripts associated with this gene.

Gene
UQCC3
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085372.2 348 Missense Mutation AGC,GGC S86G NP_001078841.1

View Full Product Details