Product Details

SNP ID
rs200833145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58267476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAAGTGGACACAAAGAGTCAAT[A/G]TCATGAGGAGAGGGTACTGCAACGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10W1 PubMed Links

Gene Details

Gene
OR10W1
Gene Name
olfactory receptor family 10 subfamily W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207374.3 785 Missense Mutation ACA,ATA T128I NP_997257.2

View Full Product Details