Product Details

SNP ID
rs200331489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4545251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTCTGGCTCACTGGCATCCCA[A/G]GGCTGGAGTCCCTACACGTCTGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52M1 PubMed Links

Gene Details

Gene
OR52M1
Gene Name
olfactory receptor family 52 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004137.1 61 Missense Mutation AGG,GGG R21G NP_001004137.1

View Full Product Details