Product Details

SNP ID
rs200014909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58507633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAGAGTCTGAAGGTGCCTGCT[A/G]CATGGATAGAGGCATTGAGGAAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5B21 PubMed Links

Gene Details

Gene
OR5B21
Gene Name
olfactory receptor family 5 subfamily B member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005218.1 473 Missense Mutation GCA,GTA A158V NP_001005218.1

View Full Product Details