Product Details

SNP ID
rs200288910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62775884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGCTGAGCGATGAGGTGGCGG[C/T]GCTGCTCGCAGAGGACGTGTGCTAT
Phenotype
MIM: 602013 MIM: 602946
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR2G PubMed Links

Gene Details

Gene
POLR2G
Gene Name
RNA polymerase II subunit G
There are no transcripts associated with this gene.

Gene
TAF6L
Gene Name
TATA-box binding protein associated factor 6 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006473.3 194 Missense Mutation GCG,GTG A34V NP_006464.1
XM_005273714.1 194 Missense Mutation GCG,GTG A34V XP_005273771.1
XM_017017100.1 194 Intron XP_016872589.1

View Full Product Details