Product Details

SNP ID
rs200130416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7000062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGGACCCTCTGATGAATGTGAA[G/T]ATTAGAACTGTGACTAAAACCCTTT
Phenotype
MIM: 605015
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF214 PubMed Links

Gene Details

Gene
ZNF214
Gene Name
zinc finger protein 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013249.2 1999 Missense Mutation ATT,CTT I541L NP_037381.2
XM_005253128.3 1999 Missense Mutation ATT,CTT I541L XP_005253185.1
XM_005253129.3 1999 Missense Mutation ATT,CTT I527L XP_005253186.1
XM_006718308.3 1999 Missense Mutation ATT,CTT I541L XP_006718371.1
XM_006718309.3 1999 Missense Mutation ATT,CTT I527L XP_006718372.1
XM_011520355.2 1999 Missense Mutation ATT,CTT I552L XP_011518657.1
XM_011520357.2 1999 Missense Mutation ATT,CTT I527L XP_011518659.1
XM_017018269.1 1999 Missense Mutation ATT,CTT I527L XP_016873758.1

View Full Product Details