Product Details

SNP ID
rs200110526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123939872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACCTAGGATGCTGGTTGACTTG[C/G]TCTCAGGCAACCCTACCATTTCCTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4D5 PubMed Links

Gene Details

Gene
OR4D5
Gene Name
olfactory receptor family 4 subfamily D member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001965.1 256 Missense Mutation CTC,GTC L86V NP_001001965.1
Gene
OR6T1
Gene Name
olfactory receptor family 6 subfamily T member 1
There are no transcripts associated with this gene.

View Full Product Details