Product Details

SNP ID
rs200023997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4386202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCAGCCCTGGCACATGGCACA[C/T]ACTCCTGAGTTCTGGAGAGGTAATA
Phenotype
MIM: 109092
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52B3P PubMed Links

Gene Details

Gene
OR52B3P
Gene Name
olfactory receptor family 52 subfamily B member 3 pseudogene
There are no transcripts associated with this gene.

Gene
TRIM21
Gene Name
tripartite motif containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003141.3 927 Missense Mutation ATG,GTG M272V NP_003132.2

View Full Product Details