Product Details

SNP ID
rs200094851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72228461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCTTCTTCCTGTAGAGGGTGA[A/G]CGAGAGCCGGACCCACCGGATGACC
Phenotype
MIM: 136425 MIM: 600829
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLR2 PubMed Links

Gene Details

Gene
FOLR2
Gene Name
folate receptor beta
There are no transcripts associated with this gene.

Gene
INPPL1
Gene Name
inositol polyphosphate phosphatase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001567.3 665 Silent Mutation GAA,GAG E120E NP_001558.3
XM_005273978.3 665 Silent Mutation GAA,GAG E120E XP_005274035.1
XM_005273979.4 665 Silent Mutation GAA,GAG E120E XP_005274036.1
XM_011544999.2 665 Silent Mutation GAA,GAG E120E XP_011543301.1
XM_011545000.2 665 Silent Mutation GAA,GAG E120E XP_011543302.1
XM_017017673.1 665 Silent Mutation GAA,GAG E110E XP_016873162.1

View Full Product Details