Product Details
- SNP ID
-
rs200097455
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59576714 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGTGCCTTATAGGGATCATATG[A/G]TGTGCCTAAAAGGAAAAGAGAGGAA
- Phenotype
-
MIM: 167040
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OSBP
PubMed Links
Gene Details
- Gene
- OSBP
- Gene Name
- oxysterol binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002556.2 |
2767 |
Missense Mutation |
CCA,TCA |
P763S |
NP_002547.1 |
View Full Product Details