Product Details
- SNP ID
-
rs200053801
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:102863100 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAAAACCATTAATTACACCATTT[C/G]TAACAACCAAACCAGCTATTGCTTT
- Phenotype
-
MIM: 601046
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MMP12
PubMed Links
Gene Details
- Gene
- MMP12
- Gene Name
- matrix metallopeptidase 12
There are no transcripts associated with this gene.
View Full Product Details