Product Details

SNP ID
rs200282189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45924520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCACACAGCCCCAGGTCTCCTG[A/G]ATCCCCAACAAGCACTACTCCGGCC
Phenotype
MIM: 609709 MIM: 603360 MIM: 608325
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LARGE2 PubMed Links

Gene Details

Gene
LARGE2
Gene Name
LARGE xylosyl- and glucuronyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300721.1 603 Nonsense Mutation TGA,TGG *169W NP_001287650.1
NM_001300722.1 603 Nonsense Mutation TGA,TGG *138W NP_001287651.1
NM_152312.4 603 Nonsense Mutation TGA,TGG *169W NP_689525.3
XM_005252787.2 603 UTR 5 XP_005252844.1
XM_006718140.1 603 Nonsense Mutation TGA,TGG *138W XP_006718203.1
XM_006718141.3 603 Nonsense Mutation TGA,TGG *169W XP_006718204.1
XM_011519886.1 603 Nonsense Mutation TGA,TGG *169W XP_011518188.1
XM_011519888.1 603 Nonsense Mutation TGA,TGG *169W XP_011518190.1
XM_011519889.1 603 Nonsense Mutation TGA,TGG *138W XP_011518191.1
XM_011519890.1 603 Nonsense Mutation TGA,TGG *169W XP_011518192.1
XM_011519891.1 603 Nonsense Mutation TGA,TGG *169W XP_011518193.1
XM_011519892.2 603 Nonsense Mutation TGA,TGG *169W XP_011518194.1
XM_011519893.1 603 Nonsense Mutation TGA,TGG *169W XP_011518195.1
XM_017017173.1 603 Nonsense Mutation TGA,TGG *169W XP_016872662.1
Gene
PEX16
Gene Name
peroxisomal biogenesis factor 16
There are no transcripts associated with this gene.

Gene
PHF21A
Gene Name
PHD finger protein 21A
There are no transcripts associated with this gene.

View Full Product Details