Product Details
- SNP ID
-
rs200676270
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:18173393 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCTTGTCGGACTGACAGGAAACG[C/T]GGTTGTGCTCTGGCTCCTGGGCTAC
- Phenotype
-
MIM: 607230
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRGPRX4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11024531] are located under a probe and SNP(s) [rs2445179] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRGPRX4
- Gene Name
- MAS related GPR family member X4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_054032.3 |
|
Intron |
|
|
NP_473373.2 |
View Full Product Details