Product Details

SNP ID
rs200382446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94129318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCTGGCGCGCCGCAGCCATGGC[C/T]ATCGCTCAACTGGCCACGGAGTACG
Phenotype
MIM: 608420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PANX1 PubMed Links

Gene Details

Gene
PANX1
Gene Name
pannexin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015368.3 194 Silent Mutation GCC,GCT A2A NP_056183.2
XM_011542734.2 194 Intron XP_011541036.1
XM_017017464.1 194 UTR 5 XP_016872953.1

View Full Product Details