Product Details
- SNP ID
-
rs200527395
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:61353023 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCTCATTAATGCCCGAAATGAC[A/G]GATGCGATGGACAGAGAGAGGATGG
- Phenotype
-
MIM: 615844
MIM: 614459
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYB561A3
PubMed Links
Gene Details
- Gene
- CYB561A3
- Gene Name
- cytochrome b561 family member A3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001161452.1 |
1357 |
Silent Mutation |
TCC,TCT |
S170S |
NP_001154924.1 |
NM_001161454.1 |
1357 |
Silent Mutation |
TCC,TCT |
S187S |
NP_001154926.1 |
NM_001300763.1 |
1357 |
Silent Mutation |
TCC,TCT |
S170S |
NP_001287692.1 |
NM_153611.4 |
1357 |
Silent Mutation |
TCC,TCT |
S170S |
NP_705839.3 |
XM_011544821.1 |
1357 |
Silent Mutation |
TCC,TCT |
S187S |
XP_011543123.1 |
XM_011544823.1 |
1357 |
Silent Mutation |
TCC,TCT |
S170S |
XP_011543125.1 |
XM_011544824.1 |
1357 |
Missense Mutation |
CGT,TGT |
R207C |
XP_011543126.1 |
XM_017017345.1 |
1357 |
Silent Mutation |
TCC,TCT |
S187S |
XP_016872834.1 |
XM_017017346.1 |
1357 |
Silent Mutation |
TCC,TCT |
S170S |
XP_016872835.1 |
XM_017017347.1 |
1357 |
Silent Mutation |
TCC,TCT |
S51S |
XP_016872836.1 |
- Gene
- TKFC
- Gene Name
- triokinase and FMN cyclase
- Gene
- TMEM138
- Gene Name
- transmembrane protein 138
There are no transcripts associated with this gene.
View Full Product Details