Product Details

SNP ID
rs200673151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67632328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGGGGCCTTACCTTTCTCCCT[A/G]TCTGTGGCACCCTTCAGCACACAGG
Phenotype
MIM: 604648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT8 PubMed Links

Gene Details

Gene
NUDT8
Gene Name
nudix hydrolase 8
There are no transcripts associated with this gene.

Gene
TBX10
Gene Name
T-box 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005995.4 1236 Silent Mutation GAC,GAT D286D NP_005986.2
XM_011544992.2 1236 Silent Mutation GAC,GAT D331D XP_011543294.1
XM_011544993.1 1236 Silent Mutation GAC,GAT D125D XP_011543295.1

View Full Product Details