Product Details

SNP ID
rs199913185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5059200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACACACCAAGACCAATCACAGAA[A/G]CAACCTTGTTGGTGAGGATGGCGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52E2 PubMed Links

Gene Details

Gene
OR52E2
Gene Name
olfactory receptor family 52 subfamily E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005164.2 Intron NP_001005164.2

View Full Product Details