Product Details

SNP ID
rs199757596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:788241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCACAGGACCAGGGACCTCGGCC[C/T]CCAGGCCCCCCACTGGCAGCCTCAT
Phenotype
MIM: 608213 MIM: 609302
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEND1 PubMed Links

Gene Details

Gene
CEND1
Gene Name
cell cycle exit and neuronal differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016564.3 515 Silent Mutation GGA,GGG G112G NP_057648.2
Gene
LOC171391
Gene Name
uncharacterized LOC171391
There are no transcripts associated with this gene.

Gene
PANO1
Gene Name
proapoptotic nucleolar protein 1
There are no transcripts associated with this gene.

Gene
SLC25A22
Gene Name
solute carrier family 25 member 22
There are no transcripts associated with this gene.

View Full Product Details