Product Details

SNP ID
rs202205802
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6891807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAAAATGCCTTGAGACTCCCCA[C/T]AGTTGACTTCATCTTGACCACAGTT
Phenotype
MIM: 608494
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR2D2 PubMed Links

Gene Details

Gene
OR2D2
Gene Name
olfactory receptor family 2 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003700.1 694 Missense Mutation ATG,GTG M232V NP_003691.1

View Full Product Details