Product Details

SNP ID
rs201569388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64318255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTGCGCCCTGAGACGCTCAGCG[A/G]GCTATATACTCGTCGGTGGGGCCGG
Phenotype
MIM: 601998 MIM: 606583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESRRA PubMed Links

Gene Details

Gene
ESRRA
Gene Name
estrogen related receptor alpha
There are no transcripts associated with this gene.

Gene
PRDX5
Gene Name
peroxiredoxin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012094.4 168 Missense Mutation AGC,GGC S14G NP_036226.1
NM_181651.2 168 Missense Mutation AGC,GGC S14G NP_857634.1
NM_181652.2 168 Missense Mutation AGC,GGC S14G NP_857635.1
XM_005273882.4 168 Missense Mutation AGC,GGC S14G XP_005273939.2
Gene
TRMT112
Gene Name
tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286082.1 168 Intron NP_001273011.1
NM_001286084.1 168 Intron NP_001273013.1
NM_016404.2 168 Intron NP_057488.1

View Full Product Details