Product Details

SNP ID
rs201602994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62422288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCTCATGGACACACCCTCCAG[C/T]TATGAGGCTGCCATGGAACTTTTCA
Phenotype
MIM: 192020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCGB1A1 PubMed Links

Gene Details

Gene
SCGB1A1
Gene Name
secretoglobin family 1A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003357.4 184 Silent Mutation AGC,AGT S41S NP_003348.1

View Full Product Details