Product Details

SNP ID
rs201704538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13010694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGGCGGCGCGGGAGGAGCCCC[C/T]GGAGCCCGACGGTGGCCCCGACGGC
Phenotype
MIM: 614713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASSF10 PubMed Links

Gene Details

Gene
RASSF10
Gene Name
Ras association domain family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080521.2 1272 Missense Mutation CCG,CTG P373L NP_001073990.2

View Full Product Details