Product Details
- SNP ID
-
rs201519706
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:94544731 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCTGTGGTGGGAGCCCTTCGGG[A/G]GGCGCGATAGCGCCCCGAGGCCGCC
- Phenotype
-
MIM: 104230
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FUT4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs553674609] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FUT4
- Gene Name
- fucosyltransferase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002033.3 |
881 |
Missense Mutation |
AGG,GGG |
R200G |
NP_002024.1 |
- Gene
- LOC105369438
- Gene Name
- uncharacterized LOC105369438
There are no transcripts associated with this gene.
View Full Product Details