Product Details

SNP ID
rs201682381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34883492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCTCAATAATGGGTACCACTAA[C/T]ATATCATCATATCTGTAAGACAAAA
Phenotype
MIM: 612491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APIP PubMed Links

Gene Details

Gene
APIP
Gene Name
APAF1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015957.3 601 Missense Mutation ATA,ATG I158M NP_057041.2
XM_011520154.2 601 Intron XP_011518456.1
XM_017017875.1 601 Intron XP_016873364.1

View Full Product Details