Product Details

SNP ID
rs201944126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124570090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAATGGGGTTCTTGTCTCCCATG[A/C]ATCCCTGCAGGCCTCCCACCCAGAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR8A1 PubMed Links

Gene Details

Gene
OR8A1
Gene Name
olfactory receptor family 8 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005194.1 22 Missense Mutation AAT,CAT N8H NP_001005194.1

View Full Product Details