Product Details

SNP ID
rs201829333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4989741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTGCAACAATTCCATCGGAAT[G/T]GGACAGACCTACTTGACATGCAGAT
Phenotype
MIM: 605470
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MMP26 PubMed Links

Gene Details

Gene
MMP26
Gene Name
matrix metallopeptidase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021801.4 214 Missense Mutation GGG,TGG G65W NP_068573.2
XM_011520219.2 214 UTR 5 XP_011518521.1
Gene
OR51L1
Gene Name
olfactory receptor family 51 subfamily L member 1
There are no transcripts associated with this gene.

View Full Product Details