Product Details

SNP ID
rs201854990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:100992425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCCCTTGACTAAAGGTTTCCC[C/T]TTAGGGTACACATTGCTATTTAACT
Phenotype
MIM: 615936
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP42 PubMed Links

Gene Details

Gene
ARHGAP42
Gene Name
Rho GTPase activating protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152432.2 346 Intron NP_689645.2
XM_011542615.2 346 UTR 3 XP_011540917.1
XM_011542616.2 346 UTR 3 XP_011540918.1
XM_011542617.2 346 UTR 3 XP_011540919.1
XM_017017237.1 346 UTR 3 XP_016872726.1
XM_017017238.1 346 UTR 3 XP_016872727.1
Gene
TMEM133
Gene Name
transmembrane protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032021.2 346 Silent Mutation CCC,CCT P39P NP_114410.1

View Full Product Details