Product Details
- SNP ID
-
rs201385605
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72004092 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGCTGAAGGCCATCGACTGGCGC[C/T]GGTCAGCCTGCAAGGAAGGGCTGTC
- Phenotype
-
MIM: 604113
MIM: 164009
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL18BP
PubMed Links
Gene Details
- Gene
- IL18BP
- Gene Name
- interleukin 18 binding protein
There are no transcripts associated with this gene.
- Gene
- NUMA1
- Gene Name
- nuclear mitotic apparatus protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286561.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2030R |
NP_001273490.1 |
NM_006185.3 |
6336 |
Missense Mutation |
CAG,CGG |
Q2044R |
NP_006176.2 |
XM_006718564.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2044R |
XP_006718627.1 |
XM_011545054.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543356.1 |
XM_011545055.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543357.1 |
XM_011545056.2 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543358.1 |
XM_011545057.2 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543359.1 |
XM_011545058.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543360.1 |
XM_011545059.2 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543361.1 |
XM_011545060.2 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543362.1 |
XM_011545061.2 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543363.1 |
XM_011545062.2 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543364.1 |
XM_011545063.2 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543365.1 |
XM_011545064.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2062R |
XP_011543366.1 |
XM_011545065.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2048R |
XP_011543367.1 |
XM_011545066.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2030R |
XP_011543368.1 |
XM_017017830.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2030R |
XP_016873319.1 |
XM_017017831.1 |
6336 |
Missense Mutation |
CAG,CGG |
Q2030R |
XP_016873320.1 |
- Gene
- RNF121
- Gene Name
- ring finger protein 121
There are no transcripts associated with this gene.
View Full Product Details