Product Details

SNP ID
rs201024769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55221103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAACCCCGAGATGCAAGTTTCC[C/T]TCTTTATTTTTTTCCTGGCCATTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10A7 PubMed Links

Gene Details

Gene
OR10A7
Gene Name
olfactory receptor family 10 subfamily A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005280.1 79 Missense Mutation CTC,TTC L27F NP_001005280.1

View Full Product Details