Product Details
- SNP ID
-
rs201028627
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:50401023 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGAGGACGACATGTTGCTTTTC[A/G]TGGAGGTGAGTGCATTATGCTAGTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM186A
PubMed Links
Gene Details
- Gene
- FAM186A
- Gene Name
- family with sequence similarity 186 member A
There are no transcripts associated with this gene.
- Gene
- LARP4
- Gene Name
- La ribonucleoprotein domain family member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001170803.1 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
NP_001164274.1 |
NM_001170804.1 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
NP_001164275.1 |
NM_001170808.1 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
NP_001164279.1 |
NM_052879.4 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
NP_443111.4 |
NM_199188.2 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
NP_954658.2 |
NM_199190.2 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
NP_954660.1 |
XM_005268613.2 |
163 |
UTR 5 |
|
|
XP_005268670.1 |
XM_011537828.2 |
163 |
UTR 5 |
|
|
XP_011536130.1 |
XM_011537829.2 |
163 |
UTR 5 |
|
|
XP_011536131.1 |
XM_011537830.2 |
163 |
Intron |
|
|
XP_011536132.1 |
XM_011537831.2 |
163 |
Intron |
|
|
XP_011536133.1 |
XM_011537833.2 |
163 |
UTR 5 |
|
|
XP_011536135.1 |
XM_011537834.2 |
163 |
UTR 5 |
|
|
XP_011536136.1 |
XM_011537835.2 |
163 |
UTR 5 |
|
|
XP_011536137.1 |
XM_011537836.2 |
163 |
UTR 5 |
|
|
XP_011536138.1 |
XM_011537838.2 |
163 |
Intron |
|
|
XP_011536140.1 |
XM_011537839.2 |
163 |
Intron |
|
|
XP_011536141.1 |
XM_011537840.2 |
163 |
UTR 5 |
|
|
XP_011536142.1 |
XM_011537841.2 |
163 |
Intron |
|
|
XP_011536143.1 |
XM_011537843.2 |
163 |
UTR 5 |
|
|
XP_011536145.1 |
XM_011537844.2 |
163 |
UTR 5 |
|
|
XP_011536146.1 |
XM_017018750.1 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
XP_016874239.1 |
XM_017018751.1 |
163 |
Intron |
|
|
XP_016874240.1 |
XM_017018752.1 |
163 |
Intron |
|
|
XP_016874241.1 |
XM_017018753.1 |
163 |
Intron |
|
|
XP_016874242.1 |
XM_017018754.1 |
163 |
UTR 5 |
|
|
XP_016874243.1 |
XM_017018755.1 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
XP_016874244.1 |
XM_017018756.1 |
163 |
UTR 5 |
|
|
XP_016874245.1 |
XM_017018757.1 |
163 |
UTR 5 |
|
|
XP_016874246.1 |
XM_017018758.1 |
163 |
UTR 5 |
|
|
XP_016874247.1 |
XM_017018759.1 |
163 |
Intron |
|
|
XP_016874248.1 |
XM_017018760.1 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
XP_016874249.1 |
XM_017018761.1 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
XP_016874250.1 |
XM_017018762.1 |
163 |
Missense Mutation |
ATG,GTG |
M5V |
XP_016874251.1 |
View Full Product Details