Product Details

SNP ID
rs200192209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5044366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTTGCCTCCGCTGCCGGACCC[A/G]GGAGTGCGGCCCTTGCCTCCGCTGC
Phenotype
MIM: 176267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNA5 PubMed Links

Gene Details

Gene
KCNA5
Gene Name
potassium voltage-gated channel subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002234.3 448 Silent Mutation CCA,CCG P73P NP_002225.2

View Full Product Details