Product Details
- SNP ID
-
rs200268233
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:95518194 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTAGACGAGGTATCAGCGTCTTCA[C/T]TGGGATGTTGGCTCCCCAACAGGAG
- Phenotype
-
MIM: 601870
MIM: 610993
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
METAP2
PubMed Links
Gene Details
- Gene
- METAP2
- Gene Name
- methionyl aminopeptidase 2
There are no transcripts associated with this gene.
- Gene
- USP44
- Gene Name
- ubiquitin specific peptidase 44
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042403.2 |
2377 |
Missense Mutation |
AAT,AGT |
N700S |
NP_001035862.1 |
NM_001278393.1 |
2377 |
Missense Mutation |
AAT,AGT |
N700S |
NP_001265322.1 |
NM_032147.4 |
2377 |
Missense Mutation |
AAT,AGT |
N700S |
NP_115523.2 |
XM_005269172.1 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_005269229.1 |
XM_005269173.1 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_005269230.1 |
XM_005269174.1 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_005269231.1 |
XM_011538800.2 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_011537102.1 |
XM_011538801.1 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_011537103.1 |
XM_011538802.1 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_011537104.1 |
XM_011538803.1 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_011537105.1 |
XM_011538804.2 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_011537106.1 |
XM_011538805.1 |
2377 |
Missense Mutation |
AAT,AGT |
N731S |
XP_011537107.1 |
XM_011538806.2 |
2377 |
Intron |
|
|
XP_011537108.1 |
XM_017020009.1 |
2377 |
Missense Mutation |
AAT,AGT |
N700S |
XP_016875498.1 |
XM_017020010.1 |
2377 |
Missense Mutation |
AAT,AGT |
N700S |
XP_016875499.1 |
XM_017020011.1 |
2377 |
Missense Mutation |
AAT,AGT |
N700S |
XP_016875500.1 |
XM_017020012.1 |
2377 |
Intron |
|
|
XP_016875501.1 |
View Full Product Details