Product Details

SNP ID
rs200518714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52362283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGCCCTCCAGCAGGCGCCTGTA[A/G]GTGGCGATCTCGATGTCCAGGCCCA
Phenotype
MIM: 602767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT85 PubMed Links

Gene Details

Gene
KRT85
Gene Name
keratin 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300810.1 832 Silent Mutation ACC,ACT T210T NP_001287739.1
NM_002283.3 832 Silent Mutation ACC,ACT T422T NP_002274.1

View Full Product Details