Product Details

SNP ID
rs200713981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112019850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTCAAGCGTCTTGCTGAAAACT[C/T]GGCTTCCAGCGATGATCTCTTGGTG
Phenotype
MIM: 602287 MIM: 612755
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERP29 PubMed Links

Gene Details

Gene
ERP29
Gene Name
endoplasmic reticulum protein 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034025.1 357 Intron NP_001029197.1
NM_006817.3 357 Missense Mutation TCG,TTG S80L NP_006808.1
XM_017018720.1 357 Intron XP_016874209.1
Gene
NAA25
Gene Name
N(alpha)-acetyltransferase 25, NatB auxiliary subunit
There are no transcripts associated with this gene.

Gene
TMEM116
Gene Name
transmembrane protein 116
There are no transcripts associated with this gene.

View Full Product Details