Product Details
- SNP ID
-
rs200761884
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:110661208 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAGTTTCCTCAACATGCCCCTGA[A/G]GTCAAGGGGGGCCCTGGGTGCGGGG
- Phenotype
-
MIM: 611227
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HVCN1
PubMed Links
Gene Details
- Gene
- HVCN1
- Gene Name
- hydrogen voltage gated channel 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040107.1 |
868 |
Missense Mutation |
CTC,TTC |
L88F |
NP_001035196.1 |
NM_001256413.1 |
868 |
Missense Mutation |
CTC,TTC |
L68F |
NP_001243342.1 |
NM_032369.3 |
868 |
Missense Mutation |
CTC,TTC |
L88F |
NP_115745.2 |
XM_005253948.2 |
868 |
Missense Mutation |
CTC,TTC |
L88F |
XP_005254005.1 |
XM_011538838.1 |
868 |
Missense Mutation |
CTC,TTC |
L135F |
XP_011537140.1 |
XM_011538839.2 |
868 |
Missense Mutation |
CTC,TTC |
L135F |
XP_011537141.1 |
XM_011538840.2 |
868 |
Missense Mutation |
CTC,TTC |
L135F |
XP_011537142.1 |
XM_011538841.2 |
868 |
Missense Mutation |
CTC,TTC |
L135F |
XP_011537143.1 |
XM_011538842.2 |
868 |
Missense Mutation |
CTC,TTC |
L135F |
XP_011537144.1 |
XM_011538844.2 |
868 |
Missense Mutation |
CTC,TTC |
L135F |
XP_011537146.1 |
XM_011538845.1 |
868 |
Missense Mutation |
CTC,TTC |
L88F |
XP_011537147.1 |
XM_011538846.2 |
868 |
Missense Mutation |
CTC,TTC |
L88F |
XP_011537148.1 |
XM_011538847.1 |
868 |
Missense Mutation |
CTC,TTC |
L88F |
XP_011537149.1 |
XM_017020026.1 |
868 |
Missense Mutation |
CTC,TTC |
L135F |
XP_016875515.1 |
XM_017020027.1 |
868 |
Missense Mutation |
CTC,TTC |
L88F |
XP_016875516.1 |
View Full Product Details