Product Details

SNP ID
rs200431075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108591772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCCCTTCTCCTCTTCCTCTGC[A/G]CCCCTGCCCTCCACCATCCTGGCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM119 PubMed Links

Gene Details

Gene
TMEM119
Gene Name
transmembrane protein 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181724.2 965 Silent Mutation GGC,GGT G204G NP_859075.2
XM_011538271.2 965 Silent Mutation GGC,GGT G204G XP_011536573.1

View Full Product Details