Product Details
- SNP ID
-
rs200742397
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:56599171 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTGGTGGTTCCGCATAGAGAGT[C/T]GCCGCCGCTTGGAGGGGGAGAGCCC
- Phenotype
-
MIM: 605682
MIM: 602387
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BAZ2A
PubMed Links
Gene Details
- Gene
- BAZ2A
- Gene Name
- bromodomain adjacent to zinc finger domain 2A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001300905.1 |
5454 |
Missense Mutation |
CAA,CGA |
Q1787R |
NP_001287834.1 |
NM_013449.3 |
5454 |
Missense Mutation |
CAA,CGA |
Q1789R |
NP_038477.2 |
XM_011537797.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1795R |
XP_011536099.1 |
XM_011537798.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1788R |
XP_011536100.1 |
XM_011537799.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1766R |
XP_011536101.1 |
XM_011537800.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1815R |
XP_011536102.1 |
XM_011537801.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1796R |
XP_011536103.1 |
XM_011537803.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1794R |
XP_011536105.1 |
XM_011537804.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1794R |
XP_011536106.1 |
XM_011537805.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1794R |
XP_011536107.1 |
XM_011537806.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1794R |
XP_011536108.1 |
XM_011537807.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1718R |
XP_011536109.1 |
XM_011537809.2 |
5454 |
Missense Mutation |
CAA,CGA |
Q1246R |
XP_011536111.1 |
XM_017018740.1 |
5454 |
Missense Mutation |
CAA,CGA |
Q1794R |
XP_016874229.1 |
XM_017018741.1 |
5454 |
Missense Mutation |
CAA,CGA |
Q1794R |
XP_016874230.1 |
XM_017018742.1 |
5454 |
Missense Mutation |
CAA,CGA |
Q1757R |
XP_016874231.1 |
- Gene
- RBMS2
- Gene Name
- RNA binding motif single stranded interacting protein 2
There are no transcripts associated with this gene.
View Full Product Details