Product Details

SNP ID
rs200770928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112944476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCACAGCGCTTCACACCAACAG[G/T]TCAGGCCGGCTCCGGCGTCAAACCC
Phenotype
MIM: 603351
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OAS3 PubMed Links

Gene Details

Gene
OAS3
Gene Name
2'-5'-oligoadenylate synthetase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006187.3 480 Missense Mutation GGT,GTT G154V NP_006178.2
XM_005253889.3 480 Missense Mutation GGT,GTT G154V XP_005253946.1
XM_017019363.1 480 Missense Mutation GGT,GTT G154V XP_016874852.1

View Full Product Details