Product Details

SNP ID
rs200604426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:124326238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACGCGGTTGGTGAGCGGCGTC[C/T]GGCGGTTGCAGTCTCCCTCCGAGTG
Phenotype
MIM: 600848
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCOR2 PubMed Links

Gene Details

Gene
NCOR2
Gene Name
nuclear receptor corepressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077261.3 7469 Missense Mutation CAG,CGG Q2383R NP_001070729.2
NM_001206654.1 7469 Missense Mutation CAG,CGG Q2429R NP_001193583.1
NM_006312.5 7469 Missense Mutation CAG,CGG Q2439R NP_006303.4

View Full Product Details