Product Details

SNP ID
rs199679912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:82358624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTGCCCACGCTGCGTGCCAAGT[C/T]GCAGGGACTGCTGCAGTTCCTGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC59 PubMed Links

Gene Details

Gene
CCDC59
Gene Name
coiled-coil domain containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014167.4 128 Intron NP_054886.2
Gene
METTL25
Gene Name
methyltransferase like 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319675.1 128 UTR 5 NP_001306604.1
NM_032230.2 128 Missense Mutation TCG,TTG S20L NP_115606.2
XM_005269186.4 128 Missense Mutation TCG,TTG S20L XP_005269243.2
XM_011538827.2 128 Missense Mutation TCG,TTG S20L XP_011537129.1
XM_011538828.2 128 Missense Mutation TCG,TTG S20L XP_011537130.1
XM_011538829.2 128 Missense Mutation TCG,TTG S20L XP_011537131.1
XM_017020020.1 128 UTR 5 XP_016875509.1
XM_017020021.1 128 UTR 5 XP_016875510.1

View Full Product Details