Product Details

SNP ID
rs199823309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64410052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCGAGGATGGATGAACAGGCTC[A/T]ATTAGGGCTAAATCCAAATGCTGAT
Phenotype
MIM: 603180
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
XPOT PubMed Links

Gene Details

Gene
XPOT
Gene Name
exportin for tRNA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007235.4 523 Missense Mutation CAA,CTA Q6L NP_009166.2
XM_017018748.1 523 Missense Mutation CAA,CTA Q6L XP_016874237.1

View Full Product Details