Product Details

SNP ID
rs199733973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102197163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGATTTAAGAAATTATGTTTGGA[A/G]CCTGTGTTCTGTAAAGAGAAGGTTG
Phenotype
MIM: 613687 MIM: 176795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARPBP PubMed Links

Gene Details

Gene
PARPBP
Gene Name
PARP1 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319988.1 333 UTR 3 NP_001306917.1
NM_001319993.1 333 UTR 3 NP_001306922.1
NM_001319994.1 333 UTR 3 NP_001306923.1
NM_001319995.1 333 Intron NP_001306924.1
NM_001319996.1 333 Intron NP_001306925.1
NM_017915.4 333 UTR 3 NP_060385.3
XM_011538509.1 333 UTR 3 XP_011536811.1
XM_011538514.2 333 UTR 3 XP_011536816.1
XM_011538515.1 333 UTR 3 XP_011536817.1
XM_017019534.1 333 UTR 3 XP_016875023.1
XM_017019535.1 333 UTR 3 XP_016875024.1
XM_017019536.1 333 UTR 3 XP_016875025.1
XM_017019537.1 333 UTR 3 XP_016875026.1
XM_017019538.1 333 UTR 3 XP_016875027.1
XM_017019539.1 333 UTR 3 XP_016875028.1
XM_017019540.1 333 UTR 3 XP_016875029.1
XM_017019541.1 333 Intron XP_016875030.1
XM_017019542.1 333 UTR 3 XP_016875031.1
XM_017019543.1 333 UTR 3 XP_016875032.1
XM_017019544.1 333 UTR 3 XP_016875033.1
Gene
PMCH
Gene Name
pro-melanin concentrating hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002674.3 333 Silent Mutation NP_002665.2
XM_017019483.1 333 Silent Mutation XP_016874972.1

View Full Product Details