Product Details

SNP ID
rs199663735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68809244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCAACATGTCTGTACCTACTGA[A/T]GGTGCTGTAACCACCTCACAGATTC
Phenotype
MIM: 164785
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130075 PubMed Links

Gene Details

Gene
LOC100130075
Gene Name
SUZ RNA binding domain containing 1 pseudogene
There are no transcripts associated with this gene.

Gene
MDM2
Gene Name
MDM2 proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145337.2 285 Silent Mutation GAA,GAT E11D NP_001138809.1
NM_001145339.2 285 Silent Mutation GAA,GAT E17D NP_001138811.1
NM_001145340.2 285 Silent Mutation GAA,GAT E11D NP_001138812.1
NM_001278462.1 285 Silent Mutation GAA,GAT E11D NP_001265391.1
NM_002392.5 285 Silent Mutation GAA,GAT E17D NP_002383.2
XM_005268872.4 285 Missense Mutation GAA,GAT E11D XP_005268929.1
XM_006719399.3 285 Intron XP_006719462.1
XM_006719400.3 285 UTR 5 XP_006719463.1

View Full Product Details