Product Details

SNP ID
rs199781690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14829445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCAATAGCTATAGGATCAGGGA[G/T]GCATTTCTTGCTGGAAGCTGTAAAA
Phenotype
MIM: 110600
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ART4 PubMed Links

Gene Details

Gene
ART4
Gene Name
ADP-ribosyltransferase 4 (Dombrock blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021071.2 1237 Missense Mutation ATC,CTC I291L NP_066549.2
Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
There are no transcripts associated with this gene.

View Full Product Details