Product Details

SNP ID
rs199677891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120464099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGCATGTGATCCTTCAGGTCCT[A/G]CCAGCTGCCTGACGGAGGAAGTCCT
Phenotype
MIM: 601562 MIM: 601943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYNLL1 PubMed Links

Gene Details

Gene
DYNLL1
Gene Name
dynein light chain LC8-type 1
There are no transcripts associated with this gene.

Gene
GATC
Gene Name
glutamyl-tRNA amidotransferase subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176818.2 Intron NP_789788.1
Gene
SRSF9
Gene Name
serine and arginine rich splicing factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003769.2 Intron NP_003760.1

View Full Product Details