Product Details

SNP ID
rs201611340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48202544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACATAGCTGAAGTGTAGAGAAAG[A/G]TGACCACAGTCAGGTGAGAGGCACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10AD1 PubMed Links

Gene Details

Gene
OR10AD1
Gene Name
olfactory receptor family 10 subfamily AD member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004134.1 749 Missense Mutation ACC,ATC T250I NP_001004134.1

View Full Product Details