Product Details

SNP ID
rs201663666
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52491556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCCTCCACCAGGTCCTGCATGC[C/T]TCTGAGCTCTGAGTCCAGGCGGCCC
Phenotype
MIM: 148041
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KRT6A PubMed Links
Additional Information
For this assay, SNP(s) [rs199613662,rs534124939] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KRT6A
Gene Name
keratin 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005554.3 930 Missense Mutation AGC,GGC S241G NP_005545.1

View Full Product Details