Product Details

SNP ID
rs201878530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113058293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGTGGGAGTGGCTGAATGAGC[C/A]CAGCCGCTGGCGGCCCTACACGGCC
Phenotype
MIM: 602582
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
DTX1 PubMed Links

Gene Details

Gene
DTX1
Gene Name
deltex E3 ubiquitin ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004416.2 1079 Missense Mutation CAC,CCC H34P NP_004407.2
XM_011538009.2 1079 Missense Mutation CAC,CCC H34P XP_011536311.1

View Full Product Details